2-Chloro-6-(trifluoromethyl)nicotinaldehyde 95%

Lieferant: Apollo Scientific

PC910896-1G PC910896-5G
APOSPC910896-1GEA 81.2 EUR
APOSPC910896-1G APOSPC910896-5G
2-Chloro-6-(trifluoromethyl)nicotinaldehyde 95%
2-Chloro-6-(trifluoromethyl)nicotinaldehyde
Formel: C₇H₃ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09907845
CAS-Nummer: 944900-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR