2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 97%

Lieferant: Apollo Scientific

PC910944-1G PC910944-250MG
APOSPC910944-1GEA 377 EUR
APOSPC910944-1G APOSPC910944-250MG
2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 97%
2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine
Formel: C₇H₄F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19705789
CAS-Nummer: 73221-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR