7-Fluoro-6-methoxy-1H-indazole 96%

Lieferant: Apollo Scientific

PC910955-1G
APOSPC910955-1GEA 364 EUR
APOSPC910955-1G
7-Fluoro-6-methoxy-1H-indazole 96%
7-Fluoro-6-methoxy-1H-indazole
Formel: C₈H₇FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD23705412
CAS-Nummer: 1427438-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR