4-Fluoro-1H-indazol-5-ol 97%

Lieferant: Apollo Scientific

PC910957-1G
APOSPC910957-1GEA 364 EUR
APOSPC910957-1G
4-Fluoro-1H-indazol-5-ol 97%
4-Fluoro-1H-indazol-5-ol
Formel: C₇H₅FN₂O
Molecular Weight: 152,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17010083
CAS-Nummer: 478838-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR