2-(Difluoromethyl)-5-nitropyridine 95%

Lieferant: Apollo Scientific

PC910983-1G PC910983-250MG
APOSPC910983-1GEA 0 EUR
APOSPC910983-1G APOSPC910983-250MG
2-(Difluoromethyl)-5-nitropyridine 95%
2-(Difluoromethyl)-5-nitropyridine
Formel: C₆H₄F₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD25476575
CAS-Nummer: 1646152-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR