7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 97%
Lieferant: Apollo Scientific
PC911168-1G
PC911168-250MG
APOSPC911168-1GEA
567
EUR
APOSPC911168-1G
APOSPC911168-250MG
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Formel:
C₁₀H₁₀F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059296 CAS-Nummer: 199678-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...