7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 97%

Lieferant: Apollo Scientific

PC911168-1G PC911168-250MG
APOSPC911168-1GEA 567 EUR
APOSPC911168-1G APOSPC911168-250MG
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Formel: C₁₀H₁₀F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08059296
CAS-Nummer: 199678-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR