3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
PC9112-25G
APOSPC9112-25GEA
558
EUR
APOSPC9112-25G
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Formel:
C₄H₂F₆O₂ Molecular Weight: 196.048 g/mol Siedepunkt: 125 °C Schmelzpunkt: 46…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00464165 CAS-Nummer: 564-10-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...