3,5-Difluoro-4-formylbenzonitrile 95%

Lieferant: Apollo Scientific

PC911293-1G
APOSPC911293-1GEA 59.6 EUR
APOSPC911293-1G
3,5-Difluoro-4-formylbenzonitrile 95%
3,5-Difluoro-4-formylbenzonitrile
Formel: C₈H₃F₂NO
Molecular Weight: 167,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457661
CAS-Nummer: 467442-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR