2-(Trifluoroacetyl)pyridine 96%
Lieferant: Apollo Scientific
PC911318-1G
PC911318-5G
PC911318-10G
APOSPC911318-1GEA
190
EUR
APOSPC911318-1G
APOSPC911318-5G
APOSPC911318-10G
2-(Trifluoroacetyl)pyridine 96%
2-(Trifluoroacetyl)pyridine
Formel:
C₇H₄F₃NO Molecular Weight: 175.108 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07380748 CAS-Nummer: 33284-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...