2-(Trifluoroacetyl)pyridine 96%

Lieferant: Apollo Scientific

PC911318-1G PC911318-5G PC911318-10G
APOSPC911318-1GEA 190 EUR
APOSPC911318-1G APOSPC911318-5G APOSPC911318-10G
2-(Trifluoroacetyl)pyridine 96%
2-(Trifluoroacetyl)pyridine
Formel: C₇H₄F₃NO
Molecular Weight: 175.108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07380748
CAS-Nummer: 33284-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR