3-Methoxy-2,4,5-trifluorbenzonitril 97%

Lieferant: Apollo Scientific

Synonyme: 2,4,5-Trifluoro-3-methoxybenzonitrile

PC911357-1G
APOSPC911357-1GEA 473 EUR
APOSPC911357-1G
3-Methoxy-2,4,5-trifluorbenzonitril 97%
3-Methoxy-2,4,5-trifluorbenzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115323
CAS-Nummer: 112811-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR