5-Fluoro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 98%

Lieferant: Apollo Scientific
Achtung

PC911382-250MG
APOSPC911382-250MGEA 115 EUR
APOSPC911382-250MG
5-Fluoro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 98%
5-Fluoro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formel: C₇H₅FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12962798
CAS-Nummer: 1190314-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR