3-Fluor-4-(4-morpholinyl)anilin 98%

Lieferant: Apollo Scientific

Synonyme: 4-(2-Fluoro-4-aminophenyl)morpholine

PC911386-100G
APOSPC911386-100GEA 62.5 EUR
APOSPC911386-100G
3-Fluor-4-(4-morpholinyl)anilin 98%
3-Fluoro-4-morpholinoaniline
Formel: C10H13FN2O
Molecular Weight: 196,2214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571270
CAS-Nummer: 93246-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR