1-(4-Fluorophenyl)-1,3-butanedione 97%

Lieferant: Apollo Scientific

Synonyme: (4-Fluorobenzoyl)acetone, 1-(4-Fluorophenyl)butane-1,3-dione

PC911392-5G
APOSPC911392-5GEA 105 EUR
APOSPC911392-5G
1-(4-Fluorophenyl)-1,3-butanedione 97%
1-(4-Fluorophenyl)-1,3-butanedione
Formel: C₁₀H₉FO₂
Molecular Weight: 180.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463139
CAS-Nummer: 29681-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR