6-Fluorgramin 97%
Lieferant: Apollo Scientific
Synonyme:
1-(6-Fluoro-1H-indol-3-yl)-N,N-dimethylmethanamine
PC9114-5G
APOSPC9114-5GEA
104
EUR
APOSPC9114-5G
6-Fluorgramin 97%
6-Fluorgramin
Formel:
C₁₁H₁₃FN₂ Molecular Weight: 192,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056918 CAS-Nummer: 343-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...