4-[(4-Fluorobenzyl)oxy]-3-nitrobenzenecarbaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC9121-1G
APOSPC9121-1GEA 147 EUR
APOSPC9121-1G
4-[(4-Fluorobenzyl)oxy]-3-nitrobenzenecarbaldehyde 97%
4-[(4-Fluorobenzyl)oxy]-3-nitrobenzenecarbaldehyde
Formel: C₁₄H₁₀FNO₄
Molecular Weight: 275,24 g/mol
Schmelzpunkt: 127…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173649
CAS-Nummer: 175136-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR