3-(Difluoromethyl)-4-fluoroaniline 95%

Lieferant: Apollo Scientific
Achtung

PC912796-100MG PC912796-250MG PC912796-1G
APOSPC912796-100MGEA 108 EUR
APOSPC912796-100MG APOSPC912796-250MG APOSPC912796-1G
3-(Difluoromethyl)-4-fluoroaniline 95%
3-(Difluoromethyl)-4-fluoroaniline
Formel: C₇H₆F₃N
Lagertemperatur: Gefrierschrank
MDL: MFCD11042620
CAS-Nummer: 445303-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR