2-Chloro-4-(trifluoromethyl)nicotinic acid 97%

Lieferant: Apollo Scientific

PC9139-1G PC9139-5G
APOSPC9139-1GEA 0 EUR
APOSPC9139-1G APOSPC9139-5G
2-Chloro-4-(trifluoromethyl)nicotinic acid 97%
2-Chloro-4-(trifluoromethyl)nicotinic acid
Formel: C₇H₃ClF₃NO₂
Schmelzpunkt: 157…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458125
CAS-Nummer: 590371-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR