2-(Trifluoromethyl)quinoline-4-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 2-(Trifluoromethyl)quinoline-4-carboxylicacid

PC9141-250MG
APOSPC9141-250MGEA 104 EUR
APOSPC9141-250MG
2-(Trifluoromethyl)quinoline-4-carboxylic acid 95+%
2-(Trifluoromethyl)quinoline-4-carboxylic acid
Formel: C₁₁H₆F₃NO₂
Molecular Weight: 241,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00835356
CAS-Nummer: 18706-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR