1-Fluor-4-(methylsulfonyl)-2-nitrobenzol
Lieferant: Apollo Scientific
|
Achtung
|
PC9145-25G
PC9145-5G
APOSPC9145-25GEA
58.2
EUR
APOSPC9145-25G
APOSPC9145-5G
1-Fluor-4-(methylsulfonyl)-2-nitrobenzol
1-Fluor-4-(methylsulfonyl)-2-nitrobenzol
Formel:
C₇H₆FNO₄S Schmelzpunkt: 148…157 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00025066 CAS-Nummer: 453-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...