4-Brom-3-(trifluormethyl)chinolin 97%

Lieferant: Apollo Scientific

PC9149-1G
APOSPC9149-1GEA 237 EUR
APOSPC9149-1G
4-Brom-3-(trifluormethyl)chinolin 97%
4-Bromo-3-(trifluoromethyl)quinoline
Formel: C₁₀H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08458128
CAS-Nummer: 590371-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR