4-Fluor-3-nitrophenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-3-nitrobenzeneboronic acid

PC9161-1G PC9161-5G
APOSPC9161-1GEA 102 EUR
APOSPC9161-1G APOSPC9161-5G
4-Fluor-3-nitrophenylboronsäure
4-Fluor-3-nitrophenylboronsäure
Formel: C₆H₅BFNO₄
Molecular Weight: 184.9176 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08277236
CAS-Nummer: 352530-22-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR