Kalium(4-fluorphenyl)trifluoroborat

Lieferant: Apollo Scientific
Achtung

PC9162-1G PC9162-5G PC9162-25G
APOSPC9162-1GEA 21.9 EUR
APOSPC9162-1G APOSPC9162-5G APOSPC9162-25G
Kalium(4-fluorphenyl)trifluoroborat
Kalium(4-fluorphenyl)trifluoroborat
Formel: C₆H₄BF₄K
Lagertemperatur: Raumtemperatur
MDL: MFCD01318170
CAS-Nummer: 192863-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR