2-Fluor-3-hydroxybenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC9214-1G
APOSPC9214-1GEA
40.6
EUR
APOSPC9214-1G
2-Fluor-3-hydroxybenzoesäure
2-Fluoro-3-hydroxybenzoic acid
Formel:
C₇H₅FO₃ Molecular Weight: 156,1100 g/mol Schmelzpunkt: 187…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862066 CAS-Nummer: 91658-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...