2-(1,1,2,3,3,3-Hexafluoropropyl)tetrahydrofuran 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC9224-25G
PC9224-5G
APOSPC9224-25GEA
607
EUR
APOSPC9224-25G
APOSPC9224-5G
2-(1,1,2,3,3,3-Hexafluoropropyl)tetrahydrofuran 95%
2-(1,1,2,3,3,3-Hexafluoropropyl)tetrahydrofuran
Formel:
C₇H₈F₆O Siedepunkt: 135…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155855 CAS-Nummer: 53005-42-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...