8-Bromo-2-(trifluoromethyl)quinoline ≥98%

Lieferant: Apollo Scientific

PC9226-100MG
APOSPC9226-100MGEA 74.1 EUR
APOSPC9226-100MG
8-Bromo-2-(trifluoromethyl)quinoline ≥98%
8-Bromo-2-(trifluoromethyl)quinoline
Formel: C₁₀H₅BrF₃N
Molecular Weight: 276,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741357
CAS-Nummer: 176722-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR