2,4-Dichloro-6-(trifluoromethyl)phenylhydrazine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC9231-1G
APOSPC9231-1GEA
30.6
EUR
APOSPC9231-1G
2,4-Dichloro-6-(trifluoromethyl)phenylhydrazine 95%
2,4-Dichloro-6-(trifluoromethyl)phenylhydrazine
Formel:
C₇H₅Cl₂F₃N₂ Schmelzpunkt: 40…43 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00174090 CAS-Nummer: 107047-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...