1-(3-Trifluoromethylphenyl)imidazoline-2-thione 98%
Lieferant: Apollo Scientific
Synonyme:
1,3-Dihydro-1-[3-(trifluoromethyl)phenyl]-2H-imidazole-2-thione
PC9237-5G
APOSPC9237-5GEA
108
EUR
APOSPC9237-5G
1-(3-Trifluoromethylphenyl)imidazoline-2-thione 98%
1-(3-Trifluoromethylphenyl)imidazoline-2-thione
Formel:
C₁₀H₇F₃N₂S Schmelzpunkt: 157…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041202 CAS-Nummer: 17452-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...