2-(Difluormethoxy)phenylessigsäure 98%

Lieferant: Apollo Scientific

PC9251-1G
APOSPC9251-1GEA 55.1 EUR
APOSPC9251-1G
2-(Difluormethoxy)phenylessigsäure 98%
2-(Difluormethoxy)phenylessigsäure
Formel: C₉H₈F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00236237
CAS-Nummer: 86867-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR