Pentafluorophenyl sulphide 95%

Lieferant: Apollo Scientific
Achtung

PC9256-1G PC9256-5G PC9256-25G
APOSPC9256-1GEA 29 EUR
APOSPC9256-1G APOSPC9256-5G APOSPC9256-25G
Pentafluorophenyl sulphide 95%
Pentafluorophenyl sulphide
Formel: C₁₂F₁₀S
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000293
CAS-Nummer: 1043-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR