Methyl-3-[2-nitro-4-(trifluoromethyl)phenoxy]thiophene-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

PC9258-1G PC9258-10G
APOSPC9258-1GEA 102 EUR
APOSPC9258-1G APOSPC9258-10G
Methyl-3-[2-nitro-4-(trifluoromethyl)phenoxy]thiophene-2-carboxylate 97%
Methyl-3-[2-nitro-4-(trifluoromethyl)phenoxy]thiophene-2-carboxylate
Molecular Weight: 306.24 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00112557
CAS-Nummer: 91041-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR