3,5-Dibrom-2-fluor-4-methylpyridin 97%
Lieferant: Apollo Scientific
Synonyme:
2-Fluoro-3,5-dibromo-4-methylpyridine
PC9264-1G
APOSPC9264-1GEA
36.4
EUR
APOSPC9264-1G
3,5-Dibrom-2-fluor-4-methylpyridin 97%
3,5-Dibromo-2-fluoro-4-methylpyridine
Formel:
C₆H₄NBr₂F Molecular Weight: 268,9100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864703 CAS-Nummer: 1000340-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...