4'-(4-Fluorbenzyloxy)-3'-nitroacetophenon 95%

Lieferant: Apollo Scientific

PC9282-1G
APOSPC9282-1GEA 132 EUR
APOSPC9282-1G
4'-(4-Fluorbenzyloxy)-3'-nitroacetophenon 95%
4'-(4-Fluorbenzyloxy)-3'-nitroacetophenon
Formel: C₁₅H₁₂FNO₄
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173872
CAS-Nummer: 175136-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR