2-Fluor-3-nitro-4-picolin 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Fluor-4-methyl-3-nitropyridin

PC9283-1G
APOSPC9283-1GEA 75.7 EUR
APOSPC9283-1G
2-Fluor-3-nitro-4-picolin 95+%
2-Fluor-3-nitro-4-picolin
Formel: C₆H₅FN₂O₂
Molecular Weight: 156,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112526
CAS-Nummer: 19346-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR