2-Chlor-3-(trifluormethyl)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC9289-5G PC9289-1G PC9289-25G
APOSPC9289-5GEA 114 EUR
APOSPC9289-5G APOSPC9289-1G APOSPC9289-25G
2-Chlor-3-(trifluormethyl)benzylbromid 97%
2-Chlor-3-(trifluormethyl)benzylbromid
Formel: C₈H₅BrClF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01631539
CAS-Nummer: 261763-22-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR