2-(4-Bromo-2-fluorophenyl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-(2-carboxycycloprop-1-yl)-2-fluorobenzene

PC9302-1G
APOSPC9302-1GEA 1020 EUR
APOSPC9302-1G
2-(4-Bromo-2-fluorophenyl)cyclopropane-1-carboxylic acid
2-(4-Bromo-2-fluorophenyl)cyclopropane-1-carboxylic acid
Formel: C₁₀H₈BrFO₂
Molecular Weight: 259,072 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12068280
CAS-Nummer: 1056010-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR