4-Bromo-3-(trifluoromethyl)benzoyl chloride 95+%

Lieferant: Apollo Scientific

Synonyme: 4-BRomo-3-(trifluoromethyl)benzoyl chloride

PC9304-1G
APOSPC9304-1GEA 418 EUR
APOSPC9304-1G
4-Bromo-3-(trifluoromethyl)benzoyl chloride 95+%
4-Bromo-3-(trifluoromethyl)benzoyl chloride
Formel: C₈H₃BrClF₃O
Molecular Weight: 287.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458132
CAS-Nummer: 914636-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR