4-(Trifluoromethyl)phenylacetyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(4-(Trifluoromethyl)phenyl)acetyl chloride

PC9309-1G
APOSPC9309-1GEA 23.2 EUR
APOSPC9309-1G
4-(Trifluoromethyl)phenylacetyl chloride 95%
4-(Trifluoromethyl)phenylacetyl chloride
Formel: C₉H₆ClF₃O
Molecular Weight: 222,6 g/mol
Siedepunkt: 94…97 °C/10mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD08458136
CAS-Nummer: 74426-51-0
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR