1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9318-1G
APOSPC9318-1GEA
209
EUR
APOSPC9318-1G
1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid 97%
1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid
Formel:
C₁₂H₁₂FNO₄ Molecular Weight: 253,23 g/mol Schmelzpunkt: 205…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202775 CAS-Nummer: 497163-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...