2-[5-(2-Fluorobenzoyl)-2-thienyl]acetonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Cyanomethyl)-5-(2-fluorobenzoyl)thiophene

PC9330-1G
APOSPC9330-1GEA 165 EUR
APOSPC9330-1G
2-[5-(2-Fluorobenzoyl)-2-thienyl]acetonitrile 97%
2-[5-(2-Fluorobenzoyl)-2-thienyl]acetonitrile
Formel: C₁₃H₈FNOS
Molecular Weight: 245,28 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664551
CAS-Nummer: 303144-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR