4,4'-Difluorobenzophenone-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-(4-fluorobenzoyl)benzoic acid

PC9338-250MG
APOSPC9338-250MGEA 140 EUR
APOSPC9338-250MG
4,4'-Difluorobenzophenone-2-carboxylic acid 97%
4,4'-Difluorobenzophenone-2-carboxylic acid
Formel: C₁₄H₈F₂O₃
Molecular Weight: 262,208 g/mol
Schmelzpunkt: 131…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01935981
CAS-Nummer: 338982-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR