(4-Fluorobenzamido)-N-(2-hydroxyethyl)acetamide 97%

Lieferant: Apollo Scientific

PC9340-100MG
APOSPC9340-100MGEA 271 EUR
APOSPC9340-100MG
(4-Fluorobenzamido)-N-(2-hydroxyethyl)acetamide 97%
(4-Fluorobenzamido)-N-(2-hydroxyethyl)acetamide
Formel: C₁₁H₁₃FN₂O₃
Schmelzpunkt: 148…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00140964
CAS-Nummer: 338419-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR