N-tert-Butyl-2-nitro-4-(trifluormethyl)anilin

Lieferant: Apollo Scientific
Achtung

PC9341-1G
APOSPC9341-1GEA 270 EUR
APOSPC9341-1G
N-tert-Butyl-2-nitro-4-(trifluormethyl)anilin
N-tert-Butyl-2-nitro-4-(trifluormethyl)anilin
Formel: C₁₁H₁₃F₃N₂O₂
Schmelzpunkt: 167…168 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00707485
CAS-Nummer: 247113-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR