3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid 97%

Lieferant: Apollo Scientific

PC9346-10G PC9346-1G PC9346-5G
APOSPC9346-10GEA 800 EUR
APOSPC9346-10G APOSPC9346-1G APOSPC9346-5G
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid 97%
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid
Formel: C₁₁H₆FNO₅S
Molecular Weight: 283.24 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00214992
CAS-Nummer: 303152-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR