3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid 97%
Lieferant: Apollo Scientific
PC9346-10G
PC9346-1G
PC9346-5G
APOSPC9346-10GEA
800
EUR
APOSPC9346-10G
APOSPC9346-1G
APOSPC9346-5G
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid 97%
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid
Formel:
C₁₁H₆FNO₅S Molecular Weight: 283.24 g/mol Schmelzpunkt: 159…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00214992 CAS-Nummer: 303152-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...