1-(4-Fluorobenzyl)-4-(2-hydroxyethyl)piperazine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9347-1G
APOSPC9347-1GEA
72.5
EUR
APOSPC9347-1G
1-(4-Fluorobenzyl)-4-(2-hydroxyethyl)piperazine 97%
1-(4-Fluorobenzyl)-4-(2-hydroxyethyl)piperazine
Formel:
C₁₃H₁₉FN₂O Schmelzpunkt: 55…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203973 CAS-Nummer: 174561-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...