4-(Trifluoromethylsulphinyl)chlorobenzene

Lieferant: Apollo Scientific

PC9351-1G
APOSPC9351-1GEA 263 EUR
APOSPC9351-1G
4-(Trifluoromethylsulphinyl)chlorobenzene
4-(Trifluoromethylsulphinyl)chlorobenzene
Formel: C₇H₄ClF₃OS
Molecular Weight: 228,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01009707
CAS-Nummer: 708-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR