5-Fluor-3-indolylessigsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC9358-1G
APOSPC9358-1GEA
270
EUR
APOSPC9358-1G
5-Fluor-3-indolylessigsäure
5-Fluor-3-indolylessigsäure
Formel:
C₁₀H₈FNO₂ Schmelzpunkt: 141 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00056921 CAS-Nummer: 443-73-2 EINECS: 207-138-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...