6-(4-Fluorophenyl)-2-(methylsulphanyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Amino-6-(4-fluorophenyl)-2-(methylthio)[1,2,4]triazolo[1,5-a]pyrimidine

PC9359-500MG
APOSPC9359-500MGEA 134 EUR
APOSPC9359-500MG
6-(4-Fluorophenyl)-2-(methylsulphanyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
6-(4-Fluorophenyl)-2-(methylsulphanyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Formel: C₁₂H₁₀FN₅S
Molecular Weight: 275,31 g/mol
Schmelzpunkt: 286…288 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138905
CAS-Nummer: 338394-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR